Globe Scientific Zipper Closure Specimen Bags, Globe Scientific Inc
Supplier: Globe Scientific
4918 4922 4929
75838-690CS 204.69 CAD
75838-690 75838-692 75838-698
Globe Scientific Zipper Closure Specimen Bags, Globe Scientific Inc
These bags are designed for the safe transportation of specimens and documentation.
  • 2 mil gauge polyethylene (PE)
  • Attached document compartment
  • Zipper closure allows repeated access to specimen
  • Imprinted with biohazard symbol
Each bag is printed with the biohazard symbol and offers checkmark boxes to indicate special handling instructions.
Certifications: Complies with OSHA and NCCLS guidelines.