Whatman™ Membrane Filters, PTFE, TE Range, Whatman products (Cytiva)
Supplier: Cytiva
10411108 10411111 10411113 10411116 10411130 10411205 10411211 10411213 10411311 10411313 10411411 10411413 10411305 10411305 10411405 10411405 10411108 10411111 10411113 10411116 10411130 10411205 10411211 10411213 10411311 10411313 10411411 10411413
89187-098PK 571.88 CAD
89187-098 89187-100 89187-102 89187-104 89187-106 89187-108 89187-110 89187-112 89187-114 89187-116 89187-118 89187-120 CA74330-434 74330-434 CA11012-914 10035-000 CA89187-098 CA89187-100 CA89187-102 CA89187-104 CA89187-106 CA89187-108 CA89187-110 CA89187-112 CA89187-114 CA89187-116 CA89187-118 CA89187-120
Whatman™ Membrane Filters, PTFE, TE Range, Whatman products (Cytiva)
Filters Membrane Filters
Whatman PTFE membrane filters from Cytiva business are chemically stable and inert, making them suitable for use with aggressive organic solvents, strong acids, and alkalis.
  • The TE range features polypropylene support material for improved strength and handling
  • Temperature resistant up to 120 °C
  • Can withstand aggressive solvents, liquids, and gases that damage other membranes
One of the major applications for the PTFE membrane is the clarification of corrosives, solvents, and aggressive fluids. This includes the important requirement in HPLC analysis for sample filtration where any solid particles can cause permanent damage to the column. The 0.5 µm pore size is normally used.
Air and gas sterilization make use of the hydrophobic characteristics of PTFE membrane filters and their ability to stop aqueous aerosols. Pore sizes of 0.2 and 0.5 µm are typically used. Sterile venting of vacuum manifolds, fermentation vessels, and filtrate tanks and containers utilize PTFE 0.2 µm membranes.