Temperature Limit Control, Safety Device, Ace Glass
Supplier: Ace Glass
12331-45 12331-15 12331-20 12331-25 12331-30 12331-35 12331-40 12331-10
75846-432EA 3782.97 CAD
75846-432 75846-436 75846-438 75846-440 75846-442 75846-444 75846-446 75846-750
Temperature Limit Control, Safety Device, Ace Glass
Controllers Temperature Controllers
Single outlet, secondary temperature control safety device for applications where temperature overshoot will cause dangerous conditions
  • Upon being tripped, an audible alarm will sound, and a manual reset will be required
Used as a power source for your in-house temperature controller, the limit control will cut power to its outlet when your upper temperature setpoint is reached.
Ordering information: Includes integral rack mounting bracket. One-year limited warranty.