dNTPs Mix 5 mm, 10 mm and 25 mm, MP Biomedicals
Supplier: MP BIOMEDICALS(FKA ICN BIO) CA
Ready to use solutions of dATP, dCTP, dGTP, dTTP.
- Improve PCR efficiency
- Lower denaturation temperatures
- Increase template lengths
The quality of nucleotides used in a PCR can significantly influence the result of the reaction
dNTP mix contains 5ųmol of each dATP, dCTP, dGTP, dTTP in an aqueous solution nuetralized to pH 7.0 at 5mM.
Application: Nucleic Acid Sequencing, Nucleic Acid Labeling, Nucleic Acid Amplification, PCR, high fidelity and long PCR, LAMP-PCR, cDNA synthesis, RT-PCR, DNA Labeling, DNA sequencing
Our manufacturing process guarantees that the dNTPs are highly purified (>99%). Contaminants can often hinder PCR and result in disappointing product yield. Achieve sensitive and consistent results with our specially prepared deoxynucleotides. Also suitable for use in cDNA synthesis, DNA sequencing and labeling procedures.