Taq-&LOAD™ PCR Mastermix, MP Biomedicals, LLC

Supplier: MP BIOMEDICALS(FKA ICN BIO) CA

11EPTAL100 11EPTAL110
CA76046-282EA 552.47 CAD
CA76046-282 CA76046-284
Taq-&LOAD™ PCR Mastermix, MP Biomedicals, LLC
Nucleic Acid Reagents End-point PCR Enzymes and Kits

Taq-&LOAD™ is a 5X concentrated PCR mastermix containing MP Biomedicals recombinant Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer.


  • Molecular Biology Grade
  • Optimized for highly efficient PCR
  • Significant savings in preparation time
  • Reduced pipetting
  • Reduced risk of errors and contamination
  • Stable at 4°C


This mastermix allows direct loading after the PCR on the agarose gel without the need of loading dye


All MP Biomedical thermostable DNA polymerases are available in ready-to-use &GO™ PCR mastermix formats. &GO™ PCR Mastermixes contain all the components for PCR with the exception of template and primers. This not only saves valuable time in the laboratory,but also reduces the risk of pipetting and reagent handling errors. Each &GO™ mix is provided 5Xconcentrated and can be used to perform 20μL or 50μL reactions. To prepare the reaction, simply add 10μL of &GO™ PCR mastermix 5XC to template and primers diluted to 40μL. For smaller reactions, use 4μL of mastermix in a 20μL reaction.


Components: Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer


The included red purple dye migrates at 400bp on a 1,2% agarose gel.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR