Accelerator Free, Class 100 Nitrile Cleanroom Glove, TechNiGlove
Supplier: Techniglove
RVCR1500 RVCR1503 RVCR1504 RVCR1505 RVCR1501 RVCR1502
75845-754CS 1121.53 CAD
75845-754 75845-748 75845-750 75845-752 75845-756 75845-758
Accelerator Free, Class 100 Nitrile Cleanroom Glove, TechNiGlove
Gloves
Rival CR's new soft modulus nitrile delivers the fit and feel of latex for the ultimate stretch and comfort.
  • 100% Nitrile
  • Powder-free
  • Accelerator-free
  • Soft modulus
Rival CR 12" ambidextrous Class 100 Nitrile gloves are accelerator-free for those with skin sensitivities. TechNiGlove products contain no natural rubber or silicone, and meet the most stringent requirements for extractables and particles.
Suitable for cleanroom, pharmaceutical, biotech, and medical device manufacturing.
Packaging: Packaged 100 gloves per double poly-sealed bag, 10 bags per case.