Potassium dihydrogen phosphate, crystallised, EMPROVE® EXPERT JPC, NF, Ph. Eur., BP for biopharmaceutical production, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

1.37039.9029 1.37039.1000 1.37039.9029
CA1.37039.5000EA 752.03 CAD
CA1.37039.5000 CA1.37039.9029 EM1.37039.1000 EM1.37039.9029
Potassium dihydrogen phosphate, crystallised, EMPROVE® EXPERT JPC, NF, Ph. Eur., BP for biopharmaceutical production, SAFC®
Potassium dihydrogen phosphate

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Corresponds to Ph. Eur., BP, JPC NF

Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: 100
Melting Pt: 253
MDL Number: MFCD00011401
CAS Number: 7778-77-0
Merck Index: 13,07744

Order Now

Specification Test Results

Assay (alkalimetric, calculated on dried substance) 98.0 - 100.5 %
Assay (alkalimetric; dried substance) ≥ 99.0 %
Identity passes test
Appearance of solution passes test
Insoluble substances ≤ 0.2 %
pH-value (2 %; water) 4.2 - 4.6
pH-value (5 %; water) 4.2 - 4.5
Carbonate passes test
Condensed phosphate passes test
Chloride (Cl) ≤ 0.009 %
Fluoride (F) ≤ 0.0010 %
Sulfate (SO₄) ≤ 0.017 %
Heavy metals (as Pb) ≤ 0.0010 %
Al (Aluminium) ≤ 0.0005 %
As (Arsenic) ≤ 0.0002 %
Fe (Iron) ≤ 0.0010 %
Na (Sodium) ≤ 0.1 %
0 passes test
Pb (Lead) ≤ 0.0004 %
Endotoxins ≤ 2.5 I.U./g
Total aerobic microbial count ≤ 100 CFU/g
Total yeast and mould count ≤ 100 CFU/g
DNases (Exo- and endonucleases) (not detectable) passes test
RNases (not detectable) passes test
Proteases (not detectable) passes test
Residual solvents (ICH Q3C) excluded by manufacturing process
Reducing substances passes test
Loss on drying (105 °C) ≤ 0.5 %
Loss on drying (130 °C) ≤ 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR