SP Wilmad-LabGlass Monier-Williams Sulfites Distilling Apparatus, SP Industries
Supplier: SP Industries
LG-6675-106 LG-6675-100 LG-6675-104 LG-6675-102
14205-176EA 2945.86 CAD
14205-176 14203-102 14207-094 14204-780
SP Wilmad-LabGlass Monier-Williams Sulfites Distilling Apparatus, SP Industries
Distillation Components Distillation Sets
Used for the determination of sulfites in food and related product, this Monier-Williams Sulfites distilling apparatus is made of borosilicate glass and has Standard Taper 24/40 outer and inner joints
The complete unit consists of a special outlet adapter with a double bend, a 300mm long Allihn condenser, a cylindrical separatory funnel, a right angle inlet adapter with a bubbler tube, a right angle hose connection adapter (LG-1702-102), and a three neck 1000mL boiling flask (LG-7330-194).