Water-Filled Boom Diverter, Justrite®
Supplier: Justrite
28450 28452 28454 28456
10861-288EA 130.82 CAD
10861-288 10861-290 10861-292 10861-294
Water-Filled Boom Diverter, Justrite®
Spill Control Materials
Temporarily divert and control fluid for easy cleanup or recovery.
  • Helps comply with EPA for stormwater regulations
  • Redirects liquids away from work areas and storm drains
Use the water filled booms in storm water management, portable washdown/pressure washing diversion, spill diversion, and equipment and personnel decontamination. Flexible PVC film construction conforms to uneven surfaces such as asphalt or concrete to form a diversionary barrier away from storm drains or as a holding area for wet vacuum operations. Features easy-open push plug for filling with standard water hose.