Pop-Up Pool, Justrite®
Supplier: Justrite
28321 28323 28325
10861-282EA 446.19 CAD
10861-282 10861-284 10861-286
Pop-Up Pool, Justrite®
Spill Control Materials
Durable, PVC-coated Pop-Up Pools contain spills from punctured saddle tanks, or leaking drums, hydraulic lines, pipes, and machinery.
  • Folds tightly and stows easily
  • Embedded foam ring top raises containment wall as fluid level increases
  • Yellow color provides visibility and easy identification
  • Vinyl fabric, space-saving and lightweight
The durable PVC coated vinyl fabric space-saving and lightweight pools to contain spills from punctured saddle tanks, or leaking drums, hydraulic lines, pipes, and machinery.