Water Filled Drain Cover, Justrite®
Supplier: Justrite
28468 28472 28474
10861-272EA 278.23 CAD
10861-272 10861-276 10861-278
Water Filled Drain Cover, Justrite®
Spill Control Materials
Water Filled Drain Covers seal drains before contaminants enter nearby water systems.
  • Preventative measure to seal drains
  • Easy-open, push plug
  • Resists damage caused by UV rays
Deploy cover as a preventative measure to seal drains before washdowns, maintenance, or other cleanup tasks that may allow contaminants into nearby water systems. Features easy-open push plug for filling with standard water hose. Durable EnGuard™V, Modified PVC coated fabric construction resists damage caused by UV rays.