Ground Protection Accessories, Justrite®
Supplier: Justrite
28334 28336 28338 28340 28342 28344 28346 28348 28350 28352 28354 28356 28358 28360 28362
10861-212EA 92.22 CAD
10861-212 10861-214 10861-216 10861-218 10861-220 10861-222 10861-224 10861-226 10861-228 10861-230 10861-232 10861-234 10861-236 10861-238 10861-240
Ground Protection Accessories, Justrite®
Spill Control Materials
Improve berm longevity without sacrificing functionality
  • Ground Mats - Create a smooth, uniform deployment area with this heavy-duty woven fabric. Place under berm for improved wear resistance against rocks and construction debris.
  • Track Mats - Position inside the berm for extra protection along tire paths. Thick durable, black woven fabric protects the floor and walls from tire abrasion caused by skidding, turning, and braking.
  • Track Runners - Place this high visibility, yellow vinyl coated fabric inside the berm for moderate protection against tire abrasion.
Use these berm accessories to increase the life and performance of your berm.