KIMBLE® ULTRA-WARE® 90 mm Microfiltration Assembly with Support, DWK Life Sciences

Supplier: DWK Life Sciences (KIMBLE)
953755-0090 953770-0090
KT953755-0090CS 2366.23 CAD
KT953755-0090 KT953770-0090
KIMBLE® ULTRA-WARE® 90 mm Microfiltration Assembly with Support, DWK Life Sciences
Filters Membrane Filter Units
ULTRA-WARE® 90 mm vacuum microfiltration assemblies for large sample volumes or samples with high particulate loads that would tend to clog a 47 mm diameter filter, with fritted glass support used for general filtration.

  • Funnel is graduated from 300 to 1000 ml in 50 ml increments
  • Maximum funnel capacity is 1100 ml
  • Prefilter size is 70 mm diameter and the approximate filter area is 38.5 cm²

Ordering information: Unit consists of a coarse fritted glass support base, One liter graduated funnel, anodized aluminum clamp and a No.8 silicone stopper.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR