Maintenance Spill Berm, Justrite®
Supplier: Justrite
28400 28402 28406 28408 28410 28414 28416 28418 28422 28426
10861-174EA 1935.96 CAD
10861-174 10861-176 10861-180 10861-182 10861-184 10861-188 10861-190 10861-192 10861-196 10861-200
Maintenance Spill Berm, Justrite®
Spill Control Materials
Compact and flexible spill containment quickly contain minor spills and leaks during temporary storage of drums, machine parts, batteries, and equipment.
  • No assembly required; simply unfold and use
  • Encased urethane foam sidewalls offer easy roll-through access without the need for a ramp
  • Walls quickly spring back into place with no memory loss
Lightweight EnGuard™I, PVC coated fabric provides broad UV and chemical resistance. Anchor berm in place using the brass grommets in each corner. Anchor stakes not included. Order optional grates to keepMore Product Information