Microcaps® TLC Spotting Capillary Tubes, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
764520-0000 764500-0000 764500-0001 764500-0002 764500-0005 764500-0010 764500-0020 764500-0025 764500-0050 764500-0100
KT764520-0000CS 394.99 CAD
KT764520-0000 KT764500-0000 KT764500-0001 KT764500-0002 KT764500-0005 KT764500-0010 KT764500-0020 KT764500-0025 KT764500-0050 KT764500-0100
Microcaps® TLC Spotting Capillary Tubes, Kimble Chase
Pipettes Microcapillaries
Microcaps® precision bore glass capillary tubes are cut to predetermined lengths so that each capillary tube will hold a known fluid volume when filled.
  • Made of borosilicate glass
  • Volumetric tolerance is ±1%
  • Fluid dispensed by simple squeeze of the bulb
  • Ideal for spotting preadsorbent TLC plates
Ordering information: Supplied with one bulb assembly and one dispenser vial with 100 micropipettes.
Order Now
SPECIFICATIONS
Environmentally Preferable