PRO External Circulators with Command Touch, Lauda Brinkmann
Supplier: Lauda-Brinkmann
L000456 L000457 L000320 L000321 L000553
75840-830EA 30253.73 CAD
75840-830 75840-832 75840-844 75840-846 75840-866
PRO External Circulators with Command Touch, Lauda Brinkmann
Circulating Thermostats Closed Circulator Systems Temperature Control Systems
PRO units have advantages such as low filling volume combined with powerful Varioflex pump, resulting in rapid temperature change. Environment Friendly (GWP).
  • Large color touch screen (5.7")
  • Programmer with 100 programs; each with 50 segments
  • Weekly timer and count-down function
  • Data logging; export to USB stick
  • Individual user rights up to 20 users
  • Menu navigation in 8 languages
  • Removable touch command
  • Temperature in °C or °F
LAUDA PRO External Circulators are designed to handle the most demanding external applications
The COMMAND TOUCH controller features the highest level of customer control with a multi-touch display and upgradable software.
The low internal filling volume of the PRO permits for rapid temperature changes and dynamic control of the external process. Varying temperature ranges and heating/cooling powers ensure the optimal fit.
Certifications: CE marked.