Eisco® Transparent Turbidity Tube
Supplier: EISCO SCIENTIFIC LLC
PH1120148
470220-790EA 106.6 CAD
470220-790
Eisco® Transparent Turbidity Tube
Turbidity Meters
Hands-on field sampling tool.
  • Method of measuring turbidity in a body of water
  • Useful in environmental studies
  • Easy to read
This elegantly simple apparatus is used to determine the transparency of water in lakes, streams, oceans as well as other bodies of water. The turbidity tube is a clear tube with a small Secchi Disc of alternating black and white quadrants and provides a quick and easy way to measure turbidity, a measure of water clarity, which is in turn related to the amount of sediments or pollutants in the water.
Ordering information: Kit contains (1) 1m transparent turbidity tube, and (1) rubber stopper with drainage mechanism.