Aneroid Barometer
Supplier: United Scientific Supplies
ANBR01
470221-934EA 95.4 CAD
470221-934
Aneroid Barometer
Barometers
Instrument for measuring pressure.
  • Ring for easy hanging
  • Aligned with GLOBE protocols
Mounted on a plastic base. The aneroid barometer includes an instruction sheet.