Single Probe Thermometers with Alarm, Time Date Stamp, Thermco
Supplier: Thermco
ACC821DIG ACC821FREV ACC821INCV ACC821REFV
10025-444EA 173.01 CAD
10025-444 10025-450 10025-452 10025-454
Single Probe Thermometers with Alarm, Time Date Stamp, Thermco
Thermometers Digital Thermometers
These large digital thermometers simultaneously show the current, minimum, and maximum temperatures and update continuously.
  • Triple display
  • Visual LED and audible beeping alarms
  • Bio-safe glycol/water probe medium
  • Improved resolution to 0.1º
  • Standard size vaccine bottles
  • Min.-max. temp with time and date stamp
The probe is housed in an unbreakable plastic bottle filled with a buffering media that maintains the thermometer reading when the instrument door is opened. The thermometer and the probe bottle have dual magnets to attach to the instrument.More Product Information