Vacuum Hydrolysis Tubes, Chemglass
Supplier: Chemglass
CG-4025-01 CG-4025-04 CG-4025-02 CG-4025-03
80072-396EA 354.37 CAD
80072-396 80072-402 80072-398 80072-400
Vacuum Hydrolysis Tubes, Chemglass
Tubes Test Tubes
Vacuum hydrolysis tubes used in protein hydrolysis, amino acid separations, freeze drying or any application under vacuum.