FastPrep™ Empty Tubes, MP Biomedicals, LLC

Supplier: MP Biomedicals
115076200 115076400
CA76045-782EA 2309.26 CAD
CA76045-782 CA76045-852 CA76045-854
FastPrep™ Empty Tubes, MP Biomedicals, LLC
Tubes Culture Tubes
FastPrep® non-skirted empty tubes are used to hold samples for cell lysis and homogenization.

  • 2 ml volume
  • Hold sample for homogenization
  • Caps sold separately

Use with color-coded Caps. Add sample to empty tube, add appropriate Lysing Matrix material and lysing buffer, and apply cap. Process using FastPrep instrument or other bead-beater to lyse, grind or homogenize sample.
Order Now

SPECIFICATIONS

Application areas
Environmentally Preferable
Instrument Compatibility
Kit compatibility
Lab essential type
Material of construction
Melting point
Sample throughput
Sample type
Specific gravity
Storage temperature

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR