Sanitary Gaskets, PTFE/Elastomer Core, Chemglass
Supplier: Chemglass
CG-152-G-07 CG-152-G-10 CG-152-G-20
89492-762EA 74.98 CAD
89492-762 89492-764 89492-766
Sanitary Gaskets, PTFE/Elastomer Core, Chemglass
Seals Gaskets
Gasket grafts a PTFE envelope to an EPDM elastomer core.
  • Gasket will not creep or cold flow overtime so it maintians the integrity of its seal
  • Temperature range −73 to +177 °C