NMR Tube Cleaner, Wong, Chemglass
Supplier: Chemglass
CG-1852-01 CG-1852-02
80061-570EA 1678.84 CAD
80061-570 80061-572
NMR Tube Cleaner, Wong, Chemglass
Tubes NMR Tubes
It permits the cleaning of four NMR tubes at the same time.
  • With solvent in the reservoir, rapid suction wets the inside of the NMR tube dislodging the contamination
  • An acetone wash followed by an air stream dries the tube in seconds
  • Rate of cleaning is approximately four tubes per minute
The NMR tube is inserted into the cleaner with a poly-pro cap placed on the closed end to provide support and a vacuum seal. Lower joint is a 24/40 inner, and the hose connection has an Ext.Ø of 8.5 mm at the middle serration. Complete unit comes with an NMR tube washer and 250 ml heavy wall filter flask.