Heratherm Low Temperature Peltier Incubators, Thermo Scientific
Supplier: Thermo Fisher Scientific
51031563 51031566 51031562 51031565
75840-778EA 16748 CAD
75840-778 75840-780 75840-782 75840-784
Heratherm Low Temperature Peltier Incubators, Thermo Scientific
Incubators Cooled Incubators
Thermo Scientific Heratherm Peltier Low Temperature Incubators offer a wide temperature range and energy efficiency in two convenient sizes to meet your incubation needs!
Two sizes: benchtop model (178L/6.3 cuft) or floor standing model (381L/13.5 cuft), both size available with or without an internal outlet
These incubators boast excellent temperature uniformity and stability, the benchtop model is stackable without any tools or kit and feature a sophisticated timer function with weekly/daily or real time mode. Other convient features include a flexible shelving system for optimal space utilization and non-tip protection, small footprint per chamber size, benchtop model with leveling feet, floor model with lockable casters for easy relocation, internal glass door for limited sample disturbance and a heated door with intelligent energy control to ensure reduced condensation – even at low temperatures.
Certifications: CE, CSA.