Uvex Seismic® Safety Glasses, Honeywell Safety
Supplier: Honeywell Safety Products
S0661X S0662X S0663X
10790-548CS 14416.99 CAD
10790-548 10790-550 10790-552
Uvex Seismic® Safety Glasses, Honeywell Safety
Safety Glasses
Delivering unmatched protection against impact, wind, dust and debris.
  • Black frame
  • Clear lens
  • Uvextra anti-fog coating
Uvex Seismic offers a range of diopter strengths for workers needing increased magnification for reading, inspection and other fine detail work.
Order Now
SPECIFICATIONS
Environmentally Preferable