Protease (from Streptomyces griseus)
Supplier: MilliporeSigma
53702-10KU 53702-25KU 53702-500KU 53702-50KU
CA80601-398EA 202.1 CAD
CA80601-398 CA80601-402 CA80601-404 CA80601-406
Protease (from Streptomyces griseus)
Enzymes
A non-specific protease liquefies mucins and digests proteins to free amino acids.
  • Can be used to isolate living chondrocytes
  • Most active at neutral pH, but stable over wide range of pH and temperature
  • Lyophilized form