Eppendorf® New Brunswick™ Innova® 44/44R Stackable Incubator Shakers
Supplier: Eppendorf
M1282-0000 M1282-0010 M1282-0004 M1282-0014 M1282-0000 M1282-0004 M1282-0010 M1282-0014
75874-524EA 42823.1 CAD
75874-524 75874-530 75875-646 75875-648 CA75874-524 CA75875-646 CA75874-530 CA75875-648
Eppendorf® New Brunswick™ Innova® 44/44R Stackable Incubator Shakers
Shakers Shaking Incubators
The Innova 44 and 44R (refrigerated) are Eppendorf’s most popular shakers - they accommodate flasks up to 5 L, have the space-saving glide-up door, and the reliable Innova triple-eccentric drive. They just run forever to give you peace of mind.
  • Applicational flexibility: Accommodates flasks up to 5 L, sticky pad platform for flat bottom vessels, orbits 25 or 51 mm (1 or 2"), optional: photosynthetic light bank and humidity monitoring
  • Saves money and lab space: No internal HEPA-filter that must be replaced annually for optimal performance, 2×/3× stackable in a small footprint
  • Reliable 24/7 with high loads: Innova triple eccentric counterbalanced drive (three weight-supporting shafts; please do not confuse with 'triple counterbalanced' drives that usually come with only one weight support), max. load 26 kg/57 lbs. (solid weight test)
  • Easy cleaning: Drainable built-in spill pan with rounded corners protects the drive from accidental spills (can be used as a water reservoir to reduce sample evaporation); airflow UV-germicidal light optional
  • Easy door and platform handling: Space-saving door glides up and out of your way for easy access to all samples; slide-out platform mechanism
  • Programmability: Continuous, timed, or multi-step programming (15 steps), store up to four programs
  • Serviceability: Service drawer for quick access to all electronic and heating/cooling components without the need to move or unstack the shaker, qualified lab shaker service available
The Innova 44 and 44R (refrigerated) stackable shakers are designed for culturing a wide variety of organisms in flasks up to 5 L. The cast iron triple-eccentric counterbalanced drive is specifically designed to support heavy workloads - the design has stood the test of time for over 70 years and continues to enable scientists today. A water reservoir with drain allows for humidification of the chamber, whilst protecting the drive unit from any spills. The simple user interface allows for easy programming of speed, temperature, time and when installed, the photo-synthetic light bank. The VisioNize box can be added to enhance monitoring and adds notifications functionality.
If you need significantly higher throughput in an even smaller footprint, for example, or more advanced programming or documentation, consider the Innova S44i.
4 Stored programs.
Ordering information: Options and accessories include: UV germicidal light, humidity monitoring, three different positioning bases, documentation and monitoring with notifications with VisioNize® Lab Suite and more.