FreeZone® Tube Holders for Freeze Dryers, Labconco®
Supplier: Labconco
7442600 7442700 7379400 7379500
82010-520EA 592.5 CAD
82010-520 82010-522 75841-030 75841-032
FreeZone® Tube Holders for Freeze Dryers, Labconco®
Racks Tube Racks
Hold tubes in upright position within a flask for freeze drying.
  • Sturdy steel or aluminum construction
  • Designed to fit securely inside Fast-Freeze® and Lyph-Lock® Flasks
  • Accommodate a variety of tube sizes
Stainless steel tube holders keep test tubes in an upright position and fit inside flasks for easy transport and freeze drying
Holders for 9 each (15mL) 17 × 120 mm tubes and 4 each (50mL) 30 ×115mm tubes each fit inside a 900mL or larger flask.
Holders for 15 each 12 or 13mm tubes, and 30 each 2mL microcentrifuge tubes each fit inside a 750mLMore Product Information