CELLvo™ Matrix, StemBioSys®
Supplier: StemBioSys
CELLvo™ Matrix is a ready-to-use cell culture substrate which provides a native microenvironment that replicates the 3-dimensional "home" in which stem cells naturally reside and proliferate.
- CELLvo™ Matrix is a natural extracellular matrix synthesized in vitro by bone marrow stromal cells and includes the biochemistry and architecture normally found in the bone marrow
- CELLvo™ Matrix is an advanced stem cell culture system that replicates the 3-dimensional "home" in which stem cells naturally reside and proliferate
- Mesenchymal stem cells grown on CELLvo™ Matrix proliferate more rapidly
- Mesenchymal stem cells grown on CELLvo™ Matrix maintain a smaller cell size
- Mesenchymal stem cells grown on CELLvo™ Matrix exhibit a higher percentage of cells expressing SSEA-4
CELLvo™ Matrix is a ready-to-use cell culture substrate which provides a native microenvironment that replicates the 3-dimensional "home" in which stem cells naturally reside and proliferate.
CELLvo™ Matrix is a protein extracellular matrix that mimics the normal microenvironment of stem cells. This 3-D matrix is produced by young adult human bone marrow cells. The final product is a thin, off-white film on cell culture flasks, dishes, and substrates.
CELLvo™ Matrix is not for human use. This product should not be used for diagnostic or therapeutic purposes.
Accessories information: Accessories not included.
Delivery information: CELLvo™ Matrix arrives at room temperature but should be refrigerated at 2 to 8°C upon arrival. Avoid extended exposure to light.
Packaging: CELLvo™ Matrix is produced under aseptic conditions and should be handled using aseptic technique.
Caution: Handle CELLvo™ related products in accordance with established bio-safety practices.