Acetic acid glacial ≥100%, EMPROVE® EXPERT USP, JP, Ph. Eur., BP, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

1.37000.2500 1.37000.9010 1.37000.9025 1.37000.2500 1.37000.5000 1.37000.9010 1.37000.9025
CA1.37000.2500EA 808.44 CAD
CA1.37000.2500 CA1.37000.9010 CA1.37000.9025 EM1.37000.2500 EM1.37000.5000 EM1.37000.9010 EM1.37000.9025
Acetic acid glacial ≥100%, EMPROVE® EXPERT USP, JP, Ph. Eur., BP, SAFC®
Acetic acid

Acetic acid (glacial) 100% suitable for biopharmaceutical production EMPROVE® bio. Elemental impurity specifications have been set considering ICH Q3D (guideline for elemental impurities).


Certifications: Meets Ph. Eur., BP, JP, USP standards.


Caution: Store at +15°C to +25°C.

Formula: H₃CCOOH
Boiling Pt: 116…118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 39 °C
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2789
ADR: 8 (3),
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms Ethanoic Acid
Explosion Limit 4 - 19.9 % (V)
Ignition temperature 485 °C
pH Value 2.5 (50 g/l, H2O, 20 °C)
Vapor Pressure 15.4 hPa (20 °C)
Viscosity Kinematic 1.17 mm2/s (20 °C)
Solubility 602.9 g/l soluble
LD 50 oral LD50 Rat 3310 mg/kg
Storage Store at 15 to 25 °C
Assay (Alkalimetric) 99.8 - 100.5 %
Identity Passes Test
Colour (Hazen) ≤10 Hazen
Appearance of the substance Passes Test
Formic acid, Formates and other oxidable impurities (as HCOOH) ≤0.5 %
Titratable Base ≤0.0004 mEq/g
Solidification Temperature ≥16.3 °C
Chloride (Cl) ≤2 ppm
Sulphate (SO4) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Fe (Iron) ≤0.2 ppm
Other Residual Solvents (ICH Q3C) Excluded by Production Process
Reducing Substances (Ph Eur) Passes Test
Substances Reducing Potassium Permanganate Passes Test
Evaporation Residue ≤50 ppm
Dilution Test Passes Test
Endotoxins ≤2.5 I.U./ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR