Starch, soluble, Extra Pure pH 6.0 - 7.5 (20 g/l, H₂O, 25 °C)., SAFC®

Supplier: MilliporeSigma

Synonyms: StarchVitex® iodine indicatorAmylodextrin

1.01253.1000 1.01253.1000
EM1.01253.1000EA 1436.99 CAD
EM1.01253.1000 CA1.01253.1000
Starch, soluble, Extra Pure pH 6.0 - 7.5 (20 g/l, H₂O, 25 °C)., SAFC®
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878

Order Now

Specification Test Results

Appearance of solution (color) colourless
Appearance of solution (clearness) almost clear to slightly opalescent
Sensitivity with iodine solution passes test
Reducing substances passes test
Microbiological test passes test
Colony count (aerobic bacteria) ≤ 5000 CFU/g
Colony count (Yeasts and moulds) ≤ 500 CFU/g
Salmonella species absent in 10 g
Staphylococcus aureus absent in 1 g
Pseudomonas aeruginosa absent in 1 g
E.coli absent in 1 g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR