Air Jet Test Sieve, W.S. Tyler™
Supplier: W.S. Tyler
9410 9003
30624-364EA 998.94 CAD
30624-364 30624-362
Air Jet Test Sieve, W.S. Tyler™
Sieves Test Sieves
For use with air jet sieving systems.
  • Overall Height: 4.44 cm (13/4")
  • Depth to Cloth: 2.86 cm (11/8")
Each sieve is serialized and supplied with a “Test Sieve Certificate.” Sieves are marked with the USA Sieve Number.