Holders for Nutating Shakers, Separatory Funnel, Glas-Col
Supplier: Glas-Col
099AVH0500S 099AVH1000S 099AVH2000S
57020-291EA 1110.17 CAD
57020-291 57020-292 57020-293
Holders for Nutating Shakers, Separatory Funnel, Glas-Col
Retort Stands
Specially designed for our floor model and mid range shakers.
  • Quick release front clamp for ease in filling of funnel
  • Design eliminates excessive handling of glassware
  • Mount to shaker platform with no fasteners
These separatory funnel holders have a rear support which remains stationary while front part holding the funnel can be released and rotated 360 degrees. This permits the funnel to be filled, inverted for shaking, and then reinverted for addition and/or extraction. All while the flask remains in the holder.
Ordering information: One holder holds one funnel.
Delivery information: 7 to 10 days after order.