Holder for Nutating Shakers, Erlenmeyer Flask, Glas-Col
Supplier: Glas-Col
099AVH0250E 099AVH0500E 099AVH1000E
57020-294EA 1499.86 CAD
57020-294 57020-295 57020-296
Holder for Nutating Shakers, Erlenmeyer Flask, Glas-Col
Supports Flask Supports
Specially designed for our floor model and mid range shakers
  • Quick release front clamp for ease in filling of funnel
  • Mount to shaker platform with no fasteners
These erlenmeyer flask holders have a rear support which remains stationary while front part holding the funnel can be released and rotated 360 degrees. This permits the funnel to be filled, inverted for shaking, and then reinverted for addition and/or extraction. All while the flask remains in the holder.
Ordering information: One holder holds one flask.
Delivery information: 7-10 days after order.