Pressure Release Valve, Glas-Col
Supplier: Glas-Col
099ASST27 099ASST22
33730-292EA 831.11 CAD
33730-292 33787-636
Pressure Release Valve, Glas-Col
Valves Pressure-Relief Valves
Automatic venting of air pressure while shaking separatory funnels.
  • One way check valve
  • Maximum chemical resistive materials
  • Eliminate exploding glassware
  • Shipping weight 0.5 lbs.
The Snorkle Stopper automatically releives pressure build up of more than one pound while shaking separatory funnels. Just load the funnels onto our shaker with these in place for worry free vigorous shaking. Made of non stick ground glass backed with a stainless steel spring and PTFE closure screw.
Ordering information: One snorkle stopper included.