HelixMark® Platinum-Cured Silicone Tubing, Freudenberg Medical
Supplier: Freudenberg Medical
61-759-03 61-049-03 61-549-01 61-130-506
75831-954EA 5416.87 CAD
75831-954 75831-956 75831-960 75831-959
HelixMark® Platinum-Cured Silicone Tubing, Freudenberg Medical
Tubing Transfer Tubing
HelixMark® platinum-cured silicone tubing meets the needs of a broad range of applications and is universally applicable for the transfer of fluids under standard requirements.
  • Fully autoclavable and sterilizable
  • Ideal for use in transfer operations as well as other applications requiring non-leachable material
  • Material certification and lot traceability available
HelixMark® tubing meets USP Class VI and EP 3.1.9 biocompatiblity requirements. It is manufactured in Freudenberg Medical's FDA registered medical device facility following Good Manufacturing Practices (GMPs) in accordance with The Quality Systems Requlations.
Certifications: Materials approved in accordance with USP Class VI, ISO 10993-1, FDA 21CFR 177-2600 and EP 3.1.9 or EP 3.1.9 without appearance of solution. Meet ISO 13485 quality standards.