RAMP® Cardiac Assays, Response Biomedical
Supplier: RESPONSE BIOMEDICAL CORP
The RAMP® Platform provides healthcare professionals with a choice of cardiac markers to optimize rapid clinical diagnosis. Using a compact, easy-to-use, automated reader, the RAMP® Platform delivers objective, rapid test results with an average time to result of approximately 15 minutes.
- Liquid Quality Control available
- Approximately 15 minutes per test
- Rapid aid in diagnosis
- EDTA Whole Blood sample type
- No centrifugation required
- Standardized test procedure for all cardiac assays
The RAMP® Troponin I test is a fast, accurate and quantitative immunochromatographic test for the determination of cardiac troponin I (TnI) levels in human EDTA whole blood
The RAMP® NT-proBNP test is a fast, accurate and quantitative immunochromatographic test for the determination of N-terminal pro-brain natriuretic peptide (NT-proBNP) levels in human EDTA whole blood. The RAMP® NT-proBNP test is used in the clinical laboratory and at the point of care, where measurement of NT-proBNP aids in the diagnosis and assessment of severity in individuals suspected of having heart failure and may aid in the risk stratification of patients with heart failure.
The RAMP® Myoglobin test is a fast, accurate and quantitative immunochromatographic test for the determination of Myoglobin levels in human EDTA whole blood. Measurement of Myoglobin aids in the rapid diagnosis of acute myocardial infarction (AMI).
The RAMP® CK-MB test is a fast, accurate and quantitative immunochromatographic test for the determination of CK-MB levels in human EDTA whole blood. Measurement of CK-MB aids in the rapid diagnosis of acute myocardial infarction (AMI).
Measurement of cardiac troponin I aids in the rapid diagnosis of acute myocardial infarction (AMI).
Certifications: FDA Cleared. Health Canada Approved. CE Marked. ISO 9001 and ISO 13485 Certified.
Delivery information: RAMP Cardiac test kits shold be stored at 2-8 °C (35-46 °F) up to the stated expiration date, however are stable at 15-25 °C (59-77 °F) for 14 days. Do not freeze. RAMP Cardiac Controls shold be stored at 2-8 °C (35-46 °F) up to the stated expiration date.