RAMP® Readers, Response Biomedical
Supplier: RESPONSE BIOMEDICAL CORP
C2100 C3100
75850-412EA 7641.39 CAD
75850-412 75850-414 75851-594
RAMP® Readers, Response Biomedical
Microplate Readers
Response Biomedical Corp. delivers fast and accurate applications on the RAMP® system, with an average time to result of approximately 15 minutes. The Readers come in two options with equivalent performance: RAMP® Reader and RAMP® 200 (Control and Test Module).
  • High quality, rapid detection
  • Manufactured in North America
  • No calibration or maintenance
  • Self-diagnostic and internal quality control checks
  • Multiple access levels for enhanced security including operator lockout
  • Small footprint
  • Data can be exported to a computer or LIS
The RAMP® Reader is a fully portable, point of care testing solution
The RAMP® 200 is an advanced, multi-port reader with innovative design features and consists of a Control Module (CM) and a Test Module (TM). ​The CM provides a convenient touchscreen interface to facilitate easy menu navigation and access to patient and quality control results. The dual port TM provides high throughput capability for any facility, allowing patient tests to be run simultaneously, with the option to expand the reader system to up to three TMs (six ports).
It is a true point of care solution that is battery and/or AC power operated (runs up 100 tests on one charge).
Certifications: FDA Cleared. Health Canada Approved. CE Marked. ISO 9001 and ISO 13485 Certified.
Accessories information: The optional serial printer for the RAMP Reader is manufactured by Citizen Systems and can be configured to autoprint patient, internal quality control and liquid quality control results. The optional barcode wand is for reading barcoded sample of patient IDs, and is manufactured by Unitech. The optional label printer for the RAMP 200 is manufactured by Zebra Technologies and can be configured to autoprint patient, internal quality control and liquid quality control results. The optional hands-free, multidirectional barcode scanner is for reading barcoded patient or sample IDs, and is manufactured by Symbol Technologies.
Ordering information: For the RAMP 200, at minimum, 1 Control Module and 1 Test Module is required. Each Control Module can have up to 3 Test Modules attached.