Test Tube Racks
Supplier: Eppendorf
M1289-0002 M1289-0050 M1289-0060 M1289-0100 M1289-0200 M1289-0400 M1289-0500 M1289-0600 M1289-0001 M1289-0003 M1289-0004 M1289-0005 M1289-0006 M1289-0010 M1289-0020 M1289-0030 M1289-0040 M1289-0001 M1289-0003 M1289-0004 M1289-0005 M1289-0006 M1289-0010 M1289-0020 M1289-0030 M1289-0040 M1289-0050 M1289-0060 M1289-0100 M1289-0200 M1289-0400 M1289-0500 M1289-0600 M1289-0002
490014-802EA 747.66 CAD
490014-802 75874-780 75874-782 75874-784 75874-786 75874-788 75874-790 75874-792 75874-940 75874-942 75874-944 75874-946 75874-948 75874-950 75874-952 75874-954 75874-956 CA75874-940 CA75874-942 CA75874-944 CA75874-946 CA75874-948 CA75874-950 CA75874-952 CA75874-954 CA75874-956 CA75874-780 CA75874-782 CA75874-784 CA75874-786 CA75874-788 CA75874-790 CA75874-792 CA490014-802
Test Tube Racks
Racks Tube Racks
Made from stainless-steel with inserts to help retain tubes and prevent them from spinning in the rack.
  • Adjustable angle and tube height support
  • Stainless steel construction
Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.