Plastic Serological Pipettes, Globe Scientific
Supplier: Globe Scientific
1720 1760 1740 1700 1780 1760
76519-464BX 146.43 CAD
76519-464 76452-276 490019-702 76452-274 490019-704 490019-706
Plastic Serological Pipettes, Globe Scientific
Pipettes Serological Pipettes
Pipettes are equipped with an aerosol barrier plug that prevents both liquid and aerosols from contaminating the pipettor.
  • Non-pyrogenic and non-cytotoxic
  • RNase-free and DNase-free
  • Sterile pipettes are sterilized by gamma irradiation
  • Reverse graduations
  • Negative graduations
  • Calibrated to deliver (TD)
  • Fits all major brands of pipette controllers
These plastic serological pipettes are produced under the strictest protocols and offer the highest degree of accuracy.
These serological pipettes are produced from crystal clear polystyrene (PS) and are color-coded for easy volume identification.
Printed black graduations prevent ambiguity and guarantee precise reading with less than 2% inaccuracy at full volume. Individually wrapped and packaged in convenient dispenser boxes.