Reversed Phase, J’sphere ODS-H80, HPLC Columns, YMC AMERICA, INC
Supplier: YMC America,Inc
JH08S04-1546WT JH08S04-2546WT JH08S04-0546WT JH08S04-1046WT
97031-114EA 2324.61 CAD
97031-114 97031-130 89216-738 89216-740
Reversed Phase, J’sphere ODS-H80, HPLC Columns, YMC AMERICA, INC
Chromatography Columns
J’sphere ODS-H80 has a hydrophobic stationary phase.
  • Yields a high carbon content and providing a strong, dominant, lipophilic interaction with the nonpolar sites of the sample molecules
  • Hydrophobic stationary phase
  • Densely covered with polymeric bonded C18 groups
J’sphere ODS-H80 has a hydrophobic stationary phase. It is densely covered with polymeric bonded C18 groups yielding a high carbon content and providing a strong, dominant, lipophilic interaction with the nonpolar sites of the sample molecules. The ability to form H-bonding gives additional selectivity, which is essential for difficult separations, such as drug and corresponding metabolite discrimination.