Circular Reaction Blocks, Chemglass
Supplier: Chemglass
CG-1991-01 CG-1991-02 CG-1991-03 CG-1991-04 CG-1991-05 CG-1991-06
33501-522EA 710.73 CAD
33501-522 33501-524 33501-526 33501-528 33501-530 33501-532
Circular Reaction Blocks, Chemglass
Block Heaters Blocks
Round reaction blocks are designed to fit directly on 135 mm diameter, round top hot plate stirrers.
  • Excellent heat transfer
  • Holes spaced for optimum stirring of all vials
  • Color-anodized for durability and easy identification
Aluminum construction offers excellent heat transfer without interfering with magnetic stirring. A thermowell is provided for insertion of a temperature sensor or contact thermometer.