Hettich UNIVERSAL 320 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
1401-01 1406-01
75802-144EA 10075.39 CAD
75802-144 75802-146
Hettich UNIVERSAL 320 General Purpose Benchtop Centrifuges, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
Hettich Lab Technology's UNIVERSAL 320 general-purpose benchtop centrifuge series provides laboratories the performance and flexibility they need to perform a variety of applications.
  • Max capacity – 4 x 200 mL
  • Max RPM/RCF – 15,000/21,382
  • Temperature control (optional)
  • General-purpose
  • Cell cultures, plates, bottles, cytology
  • All-metal containment
  • Quiet operation – quiet as a normal conversation
  • Will not increase noise levels or contribute to distractions
  • Functional design - easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced safety - self-closing, auto-locking lid with steel latches
  • All-metal chamber provides reinforced security
  • Five year warranty - all Hettich centrifuges are protected by our 5-year warranty and full technical support
  • Fully certified – engineered and manufactured in accordance with all relevant safety and production standards
  • Made in Germany – German engineered and manufactured quiet, reliable and safe since 1904
Supports a comprehensive range of accessories for clinical, research, and industrial applications
Convenient application packages for conical tubes, blood tubes, cell cultures, urinalysis, plates, cytology, and shielded cytology are available. Includes everything you need to perform your application: centrifuge, rotor, buckets, and lids, where applicable.
Hettich Lab Technology is an industry-leading laboratory equipment manufacturer based in Tuttlingen, Germany. Since 1904 we have designed, engineered, and manufactured precision equipment for the modern laboratory. Known for our vast array of centrifuges and incubators, Hettich delivers on quality, safety, and reliability.
Maximum capacity of 4 x 200 mL, 40 blood or urine tubes, 32 x 15 mL conical tubes, or 10 microtiter plates. Built-to-last with Enhanced Safety for a secure workflow. Refrigerated model (UNIVERSAL 320 R) is available for temperature-sensitive samples.
Certifications: Hettich Lab Technology centrifuges are manufactured under all relevant national and international safety standards and certifications.
Ordering information: All Hettich centrifuges include a 5-year warranty. Hettich Lab Technology centrifuge packages include everything you need to complete your application: centrifuge, rotor, buckets, adapters, and lids, where applicable. Supports over 413 application configurations; please contact your Avantor representative for more information.