Ammonia solution 25%, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®

Supplier: MilliporeSigma
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

1.05422.1000 1.05422.2500 1.05422.9180
CA1.05422.1000CS 590.98 CAD
CA1.05422.1000 CA1.05422.2500 CA1.05422.9180
Ammonia solution 25%, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Ammonia solution

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Certifications: Conforms to Ph. Eur., BP

Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 37.7 °C (1013 hPa)
Melting Pt: –57.5 °C
Density: 0.903 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now

Specification Test Results

Assay (acidimetric, NH₃) 25.0 - 30.0 %
Identity passes test
Appearance of solution passes test
Density (20/20°C) 0.892 - 0.910
Carbonate (as CO₂) ≤ 0.0060 %
Chloride (Cl) ≤ 0.00010 %
Sulfate (SO₄) ≤ 0.0005 %
Pyridine and related substances ≤ 0.0002 %
Cu (Copper) ≤ 0.00005 %
Fe (Iron) ≤ 0.000025 %
Oxidizable substances passes test
Substances reducing potassium permanganate (as O) ≤ 0.00075 %
Residual solvents (ICH Q3C) ...excluded by production process
Evaporation residue ≤ 0.0018 %
Residue on ignition (as SO₄) ≤ 0.002 %
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Conforms to Ph Eur, BP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR