CELLSTAR® Cell Culture Flasks, Cell-Repellent Surface, Greiner Bio-One
Supplier: Greiner Bio-One
690985 658985 660985
10836-539CS 370.6 CAD
10836-539 10836-541 75800-006 75838-906 75838-908 75838-910 75838-912
CELLSTAR® Cell Culture Flasks, Cell-Repellent Surface, Greiner Bio-One
Flasks Cell Culture Flasks
Cell culture vessels for suspension and spheroid culture.
  • Effectively inhibits cell adhesion
  • 4 year shelf life
  • Other formats available
Greiner Bio-One introduces a new surface chemistry to effectively inhibit cell adhesion
Achieved through an innovative chemical polymer modification, Greiner Bio-One’s cell-repellent surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
Characterised by low cell attachment, the cell-repellent surface is ideal for applications such as Spheroid cultures and stem cell aggregate formation, Suspension culture of semi-adherent and adherent cells (e.g. for suspension culture of macrophages), Gel-based 3D cultures, and Magnetic 3D cell culturing (Nano3D technology).