SP Bel-Art Biohazard Disposal Bags, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
SP Bel-Art
F13164-1419_SE
470199-972PK 340.35 CAD
470199-972
SP Bel-Art Biohazard Disposal Bags, Bel-Art Products, a part of SP
Bags Waste Disposal Bags
Bright red polypropylene bags include black biohazard symbol and precautionary procedures for added safety.
  • Sterilization patch darkens when subjected to steam sterilization
  • High temperature steam autoclavable to 135 °C
Certifications: Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests.