Hettich MIKRO 220 Microliter Centrifuges, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
2200-01 2205-01 220MICRO90
75801-716EA 9352.89 CAD
75801-716 75801-718 76300-106
Hettich MIKRO 220 Microliter Centrifuges, Hettich Lab Technology
Centrifuges Microcentrifuges
The Hettich Lab Technology MIKRO 220 microliter centrifuge series offers large capacities and high-performance for high-throughput laboratories.
  • Max capacity–48 x 1.5-2.0 mL / 6 x 50 mL
  • Max RPM/RCF–18,000/31,514
  • Robust temperature control
  • 24-place 90° swing-out microliter rotor
  • For DNA, RNA and temperature-sensitive samples
  • Quiet operation–quiet as a normal conversation
  • Will not increase noise levels or contribute to distractions
  • Functional design–easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced safety–self-closing, auto-locking lid with steel latches
  • All-metal chamber provides reinforced security
  • Five year warranty–all Hettich centrifuges are protected by our 5-year warranty and full technical support
  • Fully certified –engineered and manufactured in accordance with all relevant safety and production standards
  • Made in Germany–German engineered and manufactured quiet, reliable and safe since 1904
Within a compact footprint the MIKRO 220 can run up to 48 microliter tubes per run and reaches a maximum RCF of 31,514 x G
Convenient application packages for microliter tubes, microliter strips, cyro tubes, and cell cultures are available. Includes everything you need to perform your application: centrifuge, rotor, adapters, and lids, where applicable.
Hettich Lab Technology is an industry-leading laboratory equipment manufacturer based in Tuttlingen, Germany. Since 1904 we have designed, engineered, and manufactured precision equipment for the modern laboratory. Known for our vast array of centrifuges and incubators, Hettich delivers on quality, safety, and reliability.
Also holds standard tubes up to 50 mL. Available with a robust temperature control system for temperature-sensitive samples. Ideal for molecular biology applications including PCR, DNA, and RNA. Maximum capacities of 60 microliter tubes, 12 x 5 mL conicals, 6 x 8 PCR strips, 20 cryo tubes, 6 x 15 mL tubes, and 6 x 50 mL round bottom tubes.
Certifications: Hettich Lab Technology centrifuges are manufactured under all relevant national and international safety standards and certifications.
Ordering information: All Hettich centrifuges include a 5-year warranty. Optional Hettich Lab Technology centrifuge packages are available that include everything you need to complete your application: centrifuge, rotor, buckets, adapters, and lids, where applicable. Supports over 82 application configurations; please contact your Avantor representative for more information.