Ultra-Tred ArmorCote Anti-Fatigue Mat, Wearwell®
Supplier: Wearwell
440.38X2X60BYL 440.38X2X3BYL 440.38X3X60BYL 440.38X3X5BYL 440.38X4X60BYL 440.38X3X12BYL
89020-632RL 1599.01 CAD
89020-632 89020-634 89020-636 89020-638 89020-640 89020-756
Ultra-Tred ArmorCote Anti-Fatigue Mat, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Ultra-Tred ArmorCote mats feature a more durable urethane outer skin than standard blown PVC sponge products.
  • Economic comfort for medium traffic areas
  • Urethane coating for greater abrasion resistance
  • Safety rounded edges
The tough outer skin increases service life, especially in applications where abrasion can cause the mat to fail. Ultra-Tred ArmorCote works longer where other entry-level sponge mats fail. Ideally suited for use in dry environments.