UltraSoft Tile-Top Anti-Fatigue Mat with Anti-Microbial Sponge, Wearwell®
Supplier: Wearwell
419.78X2X3AMBK 419.78X2X3AMBL 419.78X2X3AMGY 419.78X2X3AMCH 419.78X2X60AMBK 419.78X2X60AMBL 419.78X2X60AMCH 419.78X2X60AMGY 419.78X3X5AMBK 419.78X3X5AMBL 419.78X3X5AMCH 419.78X3X5AMGY 419.78X3X60AMBK 419.78X3X60AMBL 419.78X3X60AMCH 419.78X3X60AMGY 419.78X4X60AMBK 419.78X4X60AMBL 419.78X4X60AMCH 419.78X4X60AMGY
94001-018EA 395.77 CAD
94001-018 94001-020 94001-022 94001-024 94001-026 94001-028 94001-030 94001-032 94001-034 94001-036 94001-038 94001-040 94001-042 94001-044 94001-046 94001-048 94001-050 94001-052 94001-054 94001-056
UltraSoft Tile-Top Anti-Fatigue Mat with Anti-Microbial Sponge, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Combats fatigue while providing a top to bottom anti-microbial solution.
  • Premium Anti-microbial sponge
  • Non-porous, Anti-microbial surface
  • 2X the resilience and 3X the life of look-alikes
  • Maximized standing area
  • Lay-flat design
UltraSoft Tile-Top AM doubles down on combating fatigue while providing a top to bottom anti-microbial solution. An anti-microbial top surface provides top level defense and is backed with an anti-microbial Nitricell® sponge. Twice as thick, it absorbs repeated compression and bounces back to its original form better than anyMore Product Information