durAstatic® One Touch Dispensing Bottles, HDPE, Square, Menda
Supplier: Desco
35270 35271 35272 35273 35274 35275 35276 35277 35278 35282 35288 35289 35298 35299
89499-006EA 47.67 CAD
89499-006 89499-008 89499-010 89499-012 89499-014 89499-016 89499-018 89499-020 89499-022 89499-024 89499-026 89499-034 89499-036 89499-038 89499-040
durAstatic® One Touch Dispensing Bottles, HDPE, Square, Menda
Bottles Dropping Bottles
HDPE durAstatic™ bottles are dissipative and are designed to protect valuable static sensitive printed circuitry.
  • No migratory additives - reduces the chance for contamination from the bottle
  • Embossed with ESD protective symbol
  • Colored bottles for easy identification
  • Dispenses approximately 0.2 cc of liquid with each pump
In the one-touch pump, there is an opening in the center of the dish to allow the fluid to seep back down into the bottle. One-handed operation facilitates dispensing small amount of liquid into top dish. Stainless steel pump includes spring-loaded lid which stays open or stay closed minimizing evaporation and odors.